Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAFA5
Entrez Gene ID
101121083
Gene Name
TAFA chemokine like family member 5
Gene Aliases
FAM19A5
Chromosome Location
Chr.3: 239310261 - 239490373 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007052.4 XP_004007101.3 2 - 3 299 90

View Full Product Details