Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAS2R1
Entrez Gene ID
101122889
Gene Name
taste 2 receptor member 1
Gene Aliases
-
Chromosome Location
Chr.16: 69517294 - 69518184 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017090.4 XP_004017139.2 1 - 1 815 109

View Full Product Details