Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPFFR1
Entrez Gene ID
100145876
Gene Name
neuropeptide FF receptor 1
Gene Aliases
GPR147
Chromosome Location
Chr.25: 27378871 - 27400515 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962137.1 XP_027817938.1 2 - 3 213 65

View Full Product Details