Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A41
Entrez Gene ID
101105818
Gene Name
solute carrier family 25 member 41
Gene Aliases
-
Chromosome Location
Chr.5: 16554911 - 16564880 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008574.4 XP_004008623.3 6 - 7 3303 86

View Full Product Details