Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1H1orf50
Entrez Gene ID
101106120
Gene Name
chromosome 1 C1orf50 homolog
Gene Aliases
-
Chromosome Location
Chr.1: 17837308 - 17846863 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004001860.4 XP_004001909.2 3 - 4 316 79
XM_012112867.3 XP_011968257.2 3 - 4 303 79

View Full Product Details