Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYPD3
Entrez Gene ID
101107889
Gene Name
LY6/PLAUR domain containing 3
Gene Aliases
-
Chromosome Location
Chr.14: 55128839 - 55133359 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015301.4 XP_004015350.2 2 - 3 321 68

View Full Product Details