Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF3
Entrez Gene ID
101110990
Gene Name
zinc finger protein 3
Gene Aliases
-
Chromosome Location
Chr.24: 37045237 - 37054236 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021039.3 XP_004021088.1 5 - 6 590 81

View Full Product Details