Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXA10
Entrez Gene ID
101105239
Gene Name
homeobox A10
Gene Aliases
-
Chromosome Location
Chr.4: 75749595 - 75754014 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007939.3 XP_004007988.2 1 - 2 1672 53

View Full Product Details