Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MALL
Entrez Gene ID
101116299
Gene Name
mal, T cell differentiation protein like
Gene Aliases
-
Chromosome Location
Chr.3: 110884843 - 110917823 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006179.4 XP_004006228.2 2 - 3 490 83

View Full Product Details