Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTN
Entrez Gene ID
114108641
Gene Name
sentan, cilia apical structure protein
Gene Aliases
-
Chromosome Location
Chr.19: 39538525 - 39595561 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018347.4 XP_004018396.3 3 - 4 346 90

View Full Product Details