Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMR1NB
Entrez Gene ID
101117222
Gene Name
FMR1 neighbor
Gene Aliases
-
Chromosome Location
Chr.X: 94709294 - 94763512 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022287.4 XP_004022336.3 4 - 5 673 118

View Full Product Details