Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H11orf49
Entrez Gene ID
101118987
Gene Name
chromosome 15 C11orf49 homolog
Gene Aliases
-
Chromosome Location
Chr.15: 83127761 - 83340452 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979708.1 XP_027835509.1 4 - 5 467 78
XM_027979710.1 XP_027835511.1 4 - 5 467 78

View Full Product Details