Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POPDC3
Entrez Gene ID
101121428
Gene Name
popeye domain containing 3
Gene Aliases
-
Chromosome Location
Chr.8: 35432044 - 35453251 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011240.4 XP_004011289.1 3 - 4 1049 75
XM_012182709.3 XP_012038099.1 3 - 4 858 75
XM_012182710.3 XP_012038100.1 3 - 4 896 75

View Full Product Details