Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMIE
Entrez Gene ID
101116181
Gene Name
transmembrane inner ear
Gene Aliases
-
Chromosome Location
Chr.19: 54737516 - 54746210 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018507.4 XP_004018556.2 1 - 2 96 83

View Full Product Details