Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXN2
Entrez Gene ID
101106923
Gene Name
forkhead box N2
Gene Aliases
-
Chromosome Location
Chr.3: 81332297 - 81387351 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966866.1 XP_027822667.1 4 - 5 886 119

View Full Product Details