Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRG2
Entrez Gene ID
101110585
Gene Name
killer cell lectin like receptor G2
Gene Aliases
-
Chromosome Location
Chr.4: 111882561 - 111896481 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968864.1 XP_027824665.1 4 - 5 1609 108

View Full Product Details