Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEOX1
Entrez Gene ID
101110776
Gene Name
mesenchyme homeobox 1
Gene Aliases
-
Chromosome Location
Chr.11: 19489707 - 19508761 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012966.4 XP_004013015.2 1 - 2 901 77

View Full Product Details