Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG14
Entrez Gene ID
101108526
Gene Name
ALG14 UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
-
Chromosome Location
Chr.1: 76291493 - 76394130 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972245.1 XP_027828046.1 1 - 2 246 84
XM_027972247.1 XP_027828048.1 1 - 2 253 84

View Full Product Details