Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC12
Entrez Gene ID
101106672
Gene Name
zinc finger DHHC-type palmitoyltransferase 12
Gene Aliases
-
Chromosome Location
Chr.3: 7939081 - 7942581 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004023499.4 XP_004023548.2 3 - 4 340 96
XM_027966353.1 XP_027822154.1 3 - 4 333 96

View Full Product Details