Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD42
Entrez Gene ID
101106210
Gene Name
ankyrin repeat domain 42
Gene Aliases
-
Chromosome Location
Chr.21: 14051629 - 14114592 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960115.1 XP_027815916.1 3 - 4 1205 96
XM_027960117.1 XP_027815918.1 3 - 4 1208 96
XM_027960118.1 XP_027815919.1 3 - 4 1207 96
XM_027960119.1 XP_027815920.1 3 - 4 1209 96
XM_027960120.1 XP_027815921.1 2 - 3 1111 96

View Full Product Details