Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTE1
Entrez Gene ID
101117135
Gene Name
asteroid homolog 1
Gene Aliases
-
Chromosome Location
Chr.1: 281940358 - 281962573 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012103575.3 XP_011958965.1 6 - 7 2364 81

View Full Product Details