The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004022221.4 | XP_004022270.1 | 3 - 4 | 540 | 58 |
XM_012106989.3 | XP_011962379.1 | 3 - 4 | 553 | 58 |