Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR2E
Entrez Gene ID
101116826
Gene Name
RNA polymerase II, I and III subunit E
Gene Aliases
-
Chromosome Location
Chr.5: 45687389 - 45692221 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970170.1 XP_027825971.1 1 - 2 201 108
XM_027970171.1 XP_027825972.1 1 - 2 181 108

View Full Product Details