Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL43
Entrez Gene ID
101122995
Gene Name
mitochondrial ribosomal protein L43
Gene Aliases
-
Chromosome Location
Chr.22: 24440448 - 24442816 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027960591.1 XP_027816392.1 2 - 3 1096 76

View Full Product Details