Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS31
Entrez Gene ID
101121865
Gene Name
mitochondrial ribosomal protein S31
Gene Aliases
-
Chromosome Location
Chr.10: 24187704 - 24238582 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012094.4 XP_004012143.1 1 - 2 216 79
XM_027973594.1 XP_027829395.1 1 - 2 222 79
XM_027973595.1 XP_027829396.1 1 - 2 221 79

View Full Product Details