Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNGR2
Entrez Gene ID
101114289
Gene Name
synaptogyrin 2
Gene Aliases
-
Chromosome Location
Chr.11: 9283093 - 9286935 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974188.1 XP_027829989.1 2 - 3 406 75

View Full Product Details