Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
DACT3
Entrez Gene ID
101121703
Gene Name
dishevelled binding antagonist of beta catenin 3
Gene Aliases
-
Chromosome Location
Chr.14: 57255707 - 57265533 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100612.2 XP_014956098.2 3 - 4 520 81

View Full Product Details