Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARM1
Entrez Gene ID
101104239
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
-
Chromosome Location
Chr.6: 99715715 - 99845973 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012180168.3 XP_012035558.2 3 - 4 1039 62

View Full Product Details