Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6V1G1
Entrez Gene ID
101103451
Gene Name
ATPase H+ transporting V1 subunit G1
Gene Aliases
-
Chromosome Location
Chr.2: 9514182 - 9523281 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015092959.2 XP_014948445.1 2 - 3 323 87

View Full Product Details