Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TMED8
Entrez Gene ID
101110337
Gene Name
transmembrane p24 trafficking protein family member 8
Gene Aliases
-
Chromosome Location
Chr.7: 92550972 - 92583217 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015097210.2 XP_014952696.2 1 - 2 161 65

View Full Product Details