Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECHDC3
Entrez Gene ID
101108411
Gene Name
enoyl-CoA hydratase domain containing 3
Gene Aliases
-
Chromosome Location
Chr.13: 16598364 - 16632455 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099541.2 XP_014955027.2 4 - 5 787 95

View Full Product Details