Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LUZP1
Entrez Gene ID
101122671
Gene Name
leucine zipper protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 259204178 - 259305285 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005137.4 XP_004005186.2 3 - 4 3343 80
XM_012152455.3 XP_012007845.2 4 - 5 3527 80
XM_012152463.3 XP_012007853.2 4 - 5 3673 80
XM_012152481.3 XP_012007871.2 4 - 5 3959 80
XM_012152490.3 XP_012007880.2 5 - 6 3743 80
XM_015093690.2 XP_014949176.2 3 - 4 4657 80
XM_027965544.1 XP_027821345.1 5 - 6 3779 80

View Full Product Details