Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC4
Entrez Gene ID
101115548
Gene Name
exosome component 4
Gene Aliases
-
Chromosome Location
Chr.9: 14961959 - 14964725 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011604.4 XP_004011653.1 1 - 2 1111 96

View Full Product Details