Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMRTB1
Entrez Gene ID
101116952
Gene Name
DMRT like family B with proline rich C-terminal 1
Gene Aliases
-
Chromosome Location
Chr.1: 29584266 - 29591220 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012147539.3 XP_012002929.2 2 - 3 796 86

View Full Product Details