Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABARAPL1
Entrez Gene ID
101117406
Gene Name
GABA type A receptor associated protein like 1
Gene Aliases
-
Chromosome Location
Chr.3: 220062979 - 220073747 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012175401.3 XP_012030791.1 4 - 5 641 61

View Full Product Details