Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDKRB2
Entrez Gene ID
101120781
Gene Name
bradykinin receptor B2
Gene Aliases
-
Chromosome Location
Chr.18: 61625543 - 61648899 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957373.1 XP_027813174.1 1 - 2 93 89

View Full Product Details