Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIEN1
Entrez Gene ID
101103995
Gene Name
migration and invasion enhancer 1
Gene Aliases
-
Chromosome Location
Chr.11: 23090346 - 23091753 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012847.4 XP_004012896.1 1 - 2 136 68

View Full Product Details