Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR5
Entrez Gene ID
101102609
Gene Name
phospholipid scramblase family member 5
Gene Aliases
-
Chromosome Location
Chr.1: 263832946 - 263854598 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012101849.3 XP_011957239.1 3 - 4 438 75

View Full Product Details