Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHAD
Entrez Gene ID
105608408
Gene Name
chondroadherin
Gene Aliases
-
Chromosome Location
Chr.11: 27077455 - 27081693 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974710.1 XP_027830511.1 1 - 2 910 101

View Full Product Details