Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGSN
Entrez Gene ID
101102038
Gene Name
lengsin, lens protein with glutamine synthetase domain
Gene Aliases
-
Chromosome Location
Chr.2: 122661865 - 122779511 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964784.1 XP_027820585.1 3 - 4 340 140

View Full Product Details