Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHD2
Entrez Gene ID
101109897
Gene Name
EF-hand domain family member D2
Gene Aliases
-
Chromosome Location
Chr.12: 55813866 - 55831680 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014072.4 XP_004014121.4 1 - 2 370 80

View Full Product Details