Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHCHD10
Entrez Gene ID
101110091
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Gene Aliases
-
Chromosome Location
Chr.17: 80696996 - 80699373 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956832.1 XP_027812633.1 3 - 4 438 73

View Full Product Details