Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GZMM
Entrez Gene ID
101108227
Gene Name
granzyme M
Gene Aliases
-
Chromosome Location
Chr.5: 45291720 - 45299484 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096005.2 XP_014951491.1 3 - 4 1819 112

View Full Product Details