Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSC22D4
Entrez Gene ID
101104955
Gene Name
TSC22 domain family member 4
Gene Aliases
-
Chromosome Location
Chr.24: 36746695 - 36757592 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012104546.3 XP_011959936.2 3 - 4 1956 90

View Full Product Details