Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OTOS
Entrez Gene ID
101122143
Gene Name
otospiralin
Gene Aliases
-
Chromosome Location
Chr.1: 142365969 - 142366959 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965707.1 XP_027821508.1 1 - 2 86 78

View Full Product Details