Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC114116050
Entrez Gene ID
114116050
Gene Name
cyclic GMP-AMP synthase
Gene Aliases
-
Chromosome Location
Chr.8: 205908 - 231433 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972282.1 XP_027828083.1 2 - 3 911 90

Gene Symbol
CGAS
Entrez Gene ID
101117253
Gene Name
cyclic GMP-AMP synthase
Gene Aliases
MB21D1
Chromosome Location
Chr.8: 29497 - 51512 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972727.1 XP_027828528.1 2 - 3 851 90

View Full Product Details