Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC114109597
Entrez Gene ID
114109597
Gene Name
acyl-coenzyme A thioesterase 13-like
Gene Aliases
-
Chromosome Location
Chr.20: 37285499 - 37297236 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958615.1 XP_027814416.1 1 - 2 266 113

View Full Product Details