Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRSX
Entrez Gene ID
101115005
Gene Name
dehydrogenase/reductase X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 336767 - 474550 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963301.1 XP_027819102.1 5 - 6 700 52

View Full Product Details