Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAMF6
Entrez Gene ID
101120971
Gene Name
SLAM family member 6
Gene Aliases
-
Chromosome Location
Chr.1: 118382371 - 118403074 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002672.3 XP_004002721.2 5 - 6 811 85

View Full Product Details