Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP8
Entrez Gene ID
101122565
Gene Name
GTP binding protein 8 (putative)
Gene Aliases
-
Chromosome Location
Chr.1: 195356471 - 195371813 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979963.1 XP_027835764.1 2 - 3 497 93
XM_027979968.1 XP_027835769.1 2 - 3 474 93

View Full Product Details